Water Supply Standard, Turbidity, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
592 699
CA11020-142EA 343.57 CAD
CA11020-142 CA11020-144
Water Supply Standard, Turbidity, ERA (Waters)
Standards Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 1 L of this water supply, turbidity standard after dilution
Use with nephelometric methods.