Water Pollution Standard, Volatiles, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
830 710
CA11020-152EA 625.84 CAD
CA11020-152 CA11020-156
Water Pollution Standard, Volatiles, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields in excess of 200 mL of this water pollution, volatiles standard after dilution
Acetone, Acetonitrile, Acrolein, Acrylonitrile, Benzene, Bromobenzene, Bromochloromethane, Bromodichloromethane, Bromoform, Bromomethane, 2-Butanone (MEK), n-Butylbenzene, sec-Butylbenzene, tert-Butylbenzene, Carbon disulfide, Carbon tetrachloride, Chlorobenzene, Chlorodibromomethane, Chloroethane, 2-Chloroethyl vinyl ether, Chloroform, Chloromethane, 2-chlorotoluene, 4-chlorotoluene, 1,2-Dibromo-3-chloropropane, (DBCP), 1,2-Dibromoethane (EDB), Dibromomethane, 1,2-Dichlorobenzene, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, Dichlorodifluoromethane, 1,1-Dichloroethane, 1,2-Dichloroethane, cis-1,2-Dichloroethene, 1,1-Dichloroethene, trans-1,2-Dichloroethene, 1,3-Dichloropropane, 1,2-Dichloropropane, 2,2-Dichloropropane, cis-1,3-Dichloropropene, 1,1-Dichloropropene, trans-1,3-Dichloropropene, Ethylbenzene, Hexachlorobutadiene, Hexachloroethane, 2-Hexanone, Isopropylbenzene, p-Isopropyltoluene, Methyl tert-butyl ether (MTBE), 4-Methyl-2-pentanone (MIBK), Methylene chloride, Naphthalene, Nitrobenzene, n-Propylbenzene, Styrene, 1,1,1,2-Tetrachloroethane, 1,1,2,2-Tetrachloroethane, Tetrachloroethene, Toluene, 1,2,3-Trichlorobenzene, 1,2,4-Trichlorobenzene, 1,1,1-Trichloroethane, 1,1,2-Trichloroethane, Trichloroethene, Trichlorofluoromethane, 1,2,3-Trichloropropane, 1,2,4-Trimethylbenzene, 1,3,5-Trimethylbenzene, Vinyl acetate, Vinyl chloride, m&p Xylene, o-Xylene, Xylenes total.
Use with EPA methods 601, 602, 8021, 624 and 8260. This standard contains a subset of the analytes listed below at 5 to 300 µg/L.