Microbiology Standard, Wastewater Coliform MicrobE™, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
576
CA11020-158EA 557.93 CAD
CA11020-158
Microbiology Standard, Wastewater Coliform MicrobE™, ERA
Standards Environmental Analysis Standards
Includes one lyophilized quantitative standard, for use with all Clean Water Act quantitative methods
Total Coliforms, Fecal Coliforms and E. coli, are present in the range of 20-2,400 CFU/100 mL or MPN/100 mL.
Applications include MF and MPN