Soil Standard, Anions in Soil, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
543 873
CA11020-020EA 748.68 CAD
CA11020-020 CA11020-022
Soil Standard, Anions in Soil, ERA (Waters)
Standards Environmental Analysis Standards
One 40 g anions in soil, soil standard in a screw-cap bottle is designed for a DI water extraction procedure for all of the anions listed below.