Water Pollution Standard, Acids, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
834 712
CA11020-018EA 544.33 CAD
CA11020-018 CA11022-618
Water Pollution Standard, Acids, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, acids standard after dilution
Benzoic Acid, 4-Chloro-3-methylphenol, 2-Chlorophenol, 2,4-Dichlorophenol, 2,6-Dichlorophenol, 2,4-Dimethylphenol, 2,4-Dinitrophenol, 2-Methyl-4,6-dinitrophenol, 2-Methylphenol, 3 & 4-Methlyphenol, 2-Nitrophenol, 4-Nitrophenol, Pentachlorophenol, Phenol, 2,3,4,6-Tetrachlorophenol, 2,4,5-Trichlorophenol, 2,4,6-Trichlorophenol.
Use with EPA methods 604,625, 8041 and 8270. This standard contains a subset of the analytes listed below at 30 to 200 µg/L.
Order Now
SPECIFICATIONS
Environmentally Preferable