AMPLATE™ Opaque Skirted PCR Thin Wall Plates, Simport Scientific
Supplier: Simport Scientific
T324-96SKK T324-96SKW T324-384SKK T324-384SKW T324-96SKK T324-96SKW T324-384SKK T324-384SKW
CA60872-665CS 574.95 CAD
CA60872-665 CA60872-666 CA60872-667 CA60872-668 89429-068 89429-070 89429-082 89429-084
AMPLATE™ Opaque Skirted PCR Thin Wall Plates, Simport Scientific
Microplates PCR Plates
These thin wall, opaque, 96- and 384-well PCR plates are designed for chemiluminescent and fluorescent procedures.
  • Opaque 96 and 384 well PCR plates to ensure low level of background fluorescence
  • Made of virgin PP
  • Available in white or black
  • Alphanumeric grid for sample identification
  • Flat surface of wells for better sealing
  • Area for barcoding, labeling or writing
Precision molded to ensure well-to-well and plate-to-plate uniformity. Thin-walled plates are designed for rapid thermal transfer to ensure that an efficient and fast heat transfer is occurring. An alphanumeric grid helps in sample identification. To facilitate orientation, there are corner cuts on both the 96- and 384- well series.
All sealing methods can be used for oil-free operation; domed and flat cap strips (Simport T321 Series) will fit the 96 well plate.
The design of the AMPLATE™ -384 is such that each well having a 40 µl capacity can be used with reaction volumes from 2 to 30 µl capacity. Although it has 384 wells, it can be filled using automated fluid handling systems or standard multichannel pipettors.
In order to offer more surface contact between the plate and the sealing medium, such as thermal foil and adhesive sealing films, there are no cylindrical walls extending above the plate. The AMPLATE™ is skirted to allow bar coding on sides for identification and also to make it compatible with automated fluid handling systems. Holes on sides allow for precise and accurate plate positioning and removal.
Certifications: Certified RNase-, DNase-, DNA- and Pyrogen-free.
Packaging: Packed in tamperproof resealable bags of 10 plates.