Water buffered (concentrate), pillow for dilution

Supplier: Hach

2143166 2143166
CA11014-694PK 194.81 CAD
CA11014-694 104085-870
Water buffered (concentrate), pillow for dilution
Water

Buffered Dilution Water Pillows (magnesium chloride and potassium dihydrogen phosphate, makes 25 L dilution water; 1 liter per pillow).

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR