AMPLATE™ Skirted 384-Well Thin Wall PCR Plates, Simport Scientific
Supplier: Simport Scientific
T323-384SKB T323-384SKR T323-384SKY T323-384SKG T323-384SKN T323-384SKB T323-384SKG T323-384SKN T323-384SKR T323-384SKY
CA60872-661CS 800.82 CAD
CA60872-661 CA60872-663 CA60872-664 CA60872-760 CA60872-761 89429-072 89429-074 89429-076 89429-078 89429-080
AMPLATE™ Skirted 384-Well Thin Wall PCR Plates, Simport Scientific
Microplates PCR Plates
AMPLATE™ 384-well thin wall plates are precision molded to ensure well-to-well and plate-to-plate uniformity
All plates are polypropylene and autoclavable.
Each well has 40 µl capacity and can be used with reaction volumes from 2 to 30 µl. Plates are skirted to allow bar coding, and they are compatible with automated fluid handling systems. An alphanumeric grid helps in sample identification. Two corners of the plate are cut away to facilitate orientation.
Certifications: Certified RNase-, DNase-, Pyrogen-, and DNA-free.
Ordering information: Packaged in tamperproof, resealable bags of 10 plates.