Skirted AMPLATE™ 96-Well Thin Wall PCR Plates, Simport Scientific
Supplier: Simport Scientific
T323-96SKG T323-96SKN T323-96SKB T323-96SKR T323-96SKY T323-96SKB T323-96SKG T323-96SKN T323-96SKR T323-96SKY T323-96LPN
CA60872-649CS 761.66 CAD
CA60872-649 CA60872-654 CA60872-655 CA60872-657 CA60872-658 CA60872-659 CA60872-758 CA60872-759 89429-058 89429-060 89429-062 89429-064 89429-066 89429-086 89429-088 89429-090 89429-092 89429-094 89511-372
Skirted AMPLATE™ 96-Well Thin Wall PCR Plates, Simport Scientific
Microplates PCR Plates
The T323 skirted 96-well PCR plates are thin-walled and designed for rapid thermal transfer.
  • Alphanumeric grid for better identification
  • Flat surface for better sealing
  • Can be handled by robotic handling equiment
  • Area for bar coding, labeling, or writing on each side and top
  • Each well has a volume of 100 µl
  • Made of polypropylene
The skirt around the plate provides a barcoding and labeling area. They are precision-molded to ensure well-to-well and plate-to-plate uniformity and impermeable to water vapor. The Simport AMPLATE™ can be handled by roboticMore Product Information