EVAHL PAPRs (Powered Air Purifying Respirators), Complete Systems, Bullard®
Supplier: Bullard
EVAHL20LFL EVAHL20LFM EVAHL20LF2L EVAHL20LF2M EVAHL20SICHT EVAHL20SICT EVAHL20TICHT EVAHL20TICT EVAHL20TICTDB EVAHL20TJT EVAHL20TJTDB EVAHLRT3T EVAHLRT3TDB EVAHLRT4T
89402-826EA 3951.75 CAD
89402-826 89402-828 89402-830 89402-832 89402-846 89402-850 89402-854 89402-862 89402-864 89402-866 89402-868 89402-886 89402-888 89402-890
EVAHL PAPRs (Powered Air Purifying Respirators), Complete Systems, Bullard®
Respirators Respiratory Systems
The EVAHL is simple, safe, and dependable. The system features one-button operation, a choice of two air speeds, and a single filter design.
  • Certified for hazardous locations (Class I Division 2, Class II Division 2, Class III)
  • Versatile respirator for particulate or chemical environments
  • Lightweight, compact, well-balanced design
In addition, the EVAHL offers Active Flow Technology™, which maintains an airflow of 8.5 cfm (high speed), 1000 Assigned Protection Factor (depending upon headtop), and dual alarms for low flow and low battery. Finally, the EVAHL has a visual battery fuel gauge to check remaining charge at the push of a button, a brushless motor with ten times the life of a brush motor PAPR, and optional genius charger for battery trending analysis.
Certifications: NIOSH Approvals TC-21C-0922 and TC-23C-2765. ANSI/ISA-12.12.01-2007 (CSA Certificate 2510928) Class I and II, Division and Class III, Division 1 and 2 Hazardous (Classifed) Location.
Ordering information: Complete systems include blower, battery, belt, charger, headtop, breathing tube, and airflow indicator. Filters/cartridges must be ordered separately.