SP Bel-Art Automatic Self-Zeroing Burettes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37915-1010 F37915-1025 F37915-0525 F37915-0510
62866-172EA 582.24 CAD
62866-172 62866-174 62866-176 62866-178
SP Bel-Art Automatic Self-Zeroing Burettes, Bel-Art Products, a part of SP
Avoid outside contamination with this highly accurate closed system.
  • Borosilicate glass construction
  • HDPE screw cap
  • Built-in overflow
Borosilicate glass burettes have white, 0.1 mL graduations and are mounted to a high-density polyethylene screw cap
Built-in overflow automatically drains excess liquid back into reservoir. The glass stopcock has a 2 mm bore. Large, stable plastic base can be detached from reservoir. Accuracy of burette is within 1% of total volume. Graduation accuracy is Class A.
Squeezing the flexible polyethylene bottle forcesMore Product Information