Tweezers, Fine Holes in Tips, Wire Handling, Excelta Corp®
Supplier: Excelta
115-SA 116-SA 40-S 815-SA-SE 815B-SA-SE
63041-152EA 273.07 CAD
63041-152 63041-162 63042-174 89411-750 75880-650
Tweezers, Fine Holes in Tips, Wire Handling, Excelta Corp®
Forceps
These tweezers feature holes in the tips of various sizes, to hold small wires or screws firmly in place while manipulating.
  • Holds wire without damage or deformation
  • Stainless steal to prevent corrosion
These special tweezers offer grooves or holes in the tips in various sizes. They are all made of Anti-magnetic stainless steel. The 116-CF S.M.D tweezer with soft replaceable Carbofib™ straight tipped tweezer features a .039" hole in the tips. They all are ideal for handling very small wires or screws.