HyperSep™ Retain PEP SPE Cartridges
Supplier: Thermo Scientific
60107-218 60107-225
10047-164EA 780.22 CAD
10047-164 10047-168 10047-170 10047-172 10047-174 10047-176 10047-178 10047-180 10047-182 10047-186 10047-190 10047-192 10048-476 76312-776
HyperSep™ Retain PEP SPE Cartridges
Chromatography Columns
Versatile polymeric material for retention of polar and non-polar analytes.
  • High and consistent recoveries
  • Fast and easy sample preparation and method development
  • pH stable zero to 14
HyperSep™ Retain™ PEP offers reproducible and reliable sample preparation through excellent  batch-to-batch reproducibility. Many of the problems associated with traditional silica-based materials, such as inconsistent recoveries due to sorbent drying, are eliminated when using HyperSep™ Retain™ PEP. This can lead to fast sample preparation and method development.