SP Bel-Art Tapered Rear Exhaust Fume Hoods, Bel-Art Products, a part of SP
Supplier: Bel-Art
H50015-0000
16126-672EA 1310.89 CAD
16126-672
SP Bel-Art Tapered Rear Exhaust Fume Hoods, Bel-Art Products, a part of SP
Fume Hoods and Enclosures Chemical Fume Hoods
These corrosion-proof polypropylene plastic exhaust hoods are designed to rest on the rear edge of tanks and draw a stream of air across the top of the tank away from the operator.
  • Polypropylene plastic construction
  • Corrosion-proof
  • Designed to rest on rear edge of tanks
Efficiently draws a stream of air away from the operator, improving user comfort.
Material Thickness: 3/16"