THb Handheld Tissue Homogenizer, Omni International, Inc.
Supplier: Omni International
THB115
10046-846EA 3159.42 CAD
10046-846
THb Handheld Tissue Homogenizer, Omni International, Inc.
Homogenizers Handheld Homogenizers
The Omni THb Tissue Homogenizer is designed to handle a wide range of laboratory samples, from 200µL to 100mL
The THb is lightweight enough to be handheld and can also be mounted to a motor stand for lengthier processing. When used with the patented Omni Tip™ Plastic Generator Probes cross-contamination can be completely eliminated. Stainless steel generator probes are recommended for larger and tougher samples.
Note: Steel Generator Probes and Omni Tip™ Plastic Probes are sold separately.
The THb effeciently homogenizes most samples in under 30 seconds and is highly effective at cell disruption, emulsification, and particle size reduction. The THb is compatible with a wide array of generator probes for flexibility in application requirements.
Certifications: CE compliant.
Ordering information: Includes THb motor, 1-year warranty, tool kit, clamp for stand mounting, and instruction manual.