C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63508-03 63508-07
102773-636EA 407.2 CAD
102773-636 102773-644
C-Chip Disposable Hemacytometer, Electron Microscopy Sciences
Slides Counting Chambers
The world's first truly disposable Hemacytometer, the C-Chip offers precise volume control and is easy to use.
  • Quartz grade optical plastic
  • Precise design leads to highly reproducible results
  • Two or four counting chambers fitted with clearest grid pattern rich in contrast
  • Exposure to infectious material is reduced due to its closed system
  • Continuous sample containment
  • Light and unbreakable compared to glass
  • No need for cover slips
  • Time and cost saving
  • For single use
Ues for:
Blood Analysis: Blood Cell Counting
Cell Culture: Cell Concentration measurement / Cell Viability Test
Microbiology: Bacteria and Fungal Spore Counting
IVF, IUI: Sperm Counting