Oil Mist Filters, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
91004-OMF 91005-OMF 91005-RMO 91005-ROM
102773-624EA 974.13 CAD
102773-624 102773-628 102773-630 103523-950
Oil Mist Filters, Electron Microscopy Sciences
Filters Gas Filters
Protection from oil mist emissions as required by COSHH and health and safety regulations.
Mist filters capture oil mist from the outlet of pumps, which would otherwise be ejected into the atmosphere. This may happen when you use gas ballast or when you pump high gas throughputs. You can also return oil trapped in the mist filter back to the pump, although you must ensure that the process gases will not contaminate the pumpor pump oil. For the E2M1.5 use the EMF3. For the RV3, RV5, and RV8 use the EMF 10. They are very efficient at 99.999% DOP test and are also azide proof. The white bottom half of the body is semi-transparent, allowing the oil level to be monitored. Supplied with NW25 clamp, centering ring and 'O' ring, NW25 to ³/₄" BSP adaptor.
Use of optional oil return kits results in reduced oil level maintenance and savings in pump oil. If the oil element becomes blocked, an integral pressure relief valve opens.