Correlative Microscopy Coverslips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66108-02 66108-03
102773-606EA 288.29 CAD
102773-606 102773-608
Correlative Microscopy Coverslips, Electron Microscopy Sciences
Cover Glasses
EMS introduces a simple coverslip with a grid image that allows cells to grow and later be analyzed under brightfield or fluorescence microscopy before being further processed and sectioned for EM or other analysis.
  • Reticle film has been thoroughly tested with cell culture
  • Good growth of cell culture (equivalent to conventional media)
  • Good adhesion to the substrate without cell polylysine
  • Resistant to normal chemicals used in electron microscopy
  • No oxygen retention, compatible resin LR White
  • Good optical quality in brightfield & UV fluorescence
  • Excellent transparency
  • Does not deform at temperatures (positive 100C and negative liquid N2)
  • Rigid, does not float in the middle of culture
  • Easy to handle and cut with a knife or micro-punch
  • Simple sterilisation using alcohol or UV
  • Detaches easily from resin after polymerisation
  • Low cost
Applications include Light Microscopy, Fluorescence, Scanning Electron Microscopy (SEM), Transmission Electron Microscopy (TEM), and High-Pressure Cryofixation.