Counting Cells for Parasitology, Cytology and Others, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68052-23 68052-24
101765-044EA 957.2 CAD
101765-044 101765-046
Counting Cells for Parasitology, Cytology and Others, Electron Microscopy Sciences
Slides Counting Chambers
These are used to create precise volumes of liquid to enable counting of particles in a specified volume of liquid.
  • A thick slide (glass or plastic) has an area cut into its surface that will hold precise volumes of bathing fluid
  • Shallow well facilitates easy counting of contained particles
In use, a single drop of liquid is placed on the slide and a coverglass is placed on the platform providing equal and accurate subdivisions of the fluid each of which can be observed for particle counting.
Ordering information: All complete with case and 1 pair of cover glasses.