Cardboard Freezer Boxes and Dividers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61615-65 61615-70 61615-75 61615-80 61615-85
102093-720EA 144.98 CAD
102093-720 102093-722 102093-724 102093-726 102093-728 102093-730
Cardboard Freezer Boxes and Dividers, Electron Microscopy Sciences
Boxes Cooling/Freezing Boxes and Bags
For use with stainless steel freezer racks. Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes.
  • Box footprint is 133×133 mm
Select a box and a partition that ft the size and number of vials for your needs. Cardboard boxes can be labelled with Cryo-Tags for easy identification.
Packaging: Both the boxes and dividers are available in packs of 12.
Order Now
SPECIFICATIONS
Environmentally Preferable