Coverglass Staining Jars w/Plastic Caps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72242-24 72242-21
100501-230EA 456.02 CAD
100501-230 100493-696
Coverglass Staining Jars w/Plastic Caps, Electron Microscopy Sciences
Jars Staining Jars
For use with cover glass sizes 18 to 22 mm, accommodating up to eight back-to-back.
Plastic screw caps have been tested to be resistant against most staining solutions.
Order Now
SPECIFICATIONS
Environmentally Preferable