NGW2 Graticles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68025-16 68025-19 68025-21
100499-236EA 687.57 CAD
100499-236 100499-238 100499-240
NGW2 Graticles, Electron Microscopy Sciences
Graticules, Reticles and Grids
This graticule consists of a number of short lines with interruptions as long as the lines.
  • Used when making a surface to volume ratio of a structure per mass unit