Kendall Filter Pipette Tips (Barrier Tips), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72460-40 72460-05 72460-70 72460-06
100500-182CS 357.46 CAD
100500-182 100493-804 100500-188 100493-806
Kendall Filter Pipette Tips (Barrier Tips), Electron Microscopy Sciences
Pipette Tips
Kendall filter pipette tips are certified Rnase / Dnase-Free, Human DNA-Free, and PCR inhibitor-free. Their low binding surface reduces DNA and protection retention. Liquid and aerosol barrier prevent pipetter contamination.
  • Types of Membranes
  • Aerosol Barrier: Prevents aerosol contamination
  • Self-sealing Barrier (contains cellulose): Prevents aerosol and liquid contamination
  • Cellulose can contaminate samples
  • Liquid Barrier: Prevents aerosol and liquid contamination
  • Will not contaminate samples
There's a universal fit on most research-grade pipetters. CellPack keeps tips straight for multi-channel pipetters. Made from high-density polyethylene (naturally hydrophobic). Features aerosol, self-sealing and liquid barrier membranes.