Histo-Mark Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72155-01 72155-12
102097-874EA 17.41 CAD
102097-874 102097-876
Histo-Mark Pen, Electron Microscopy Sciences
Pens Ink Pens
The fine-line Histo-Mark pen is designed to work like a pen.
  • Can be used on any surface
  • Marking quality remains consistent throughout the life of the ink barrel
The Histo-Mark pen is solvent resistent, especially to alcohol, acetone, xylene, xylene subsite and toluene.