Glass Rods, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66206-20 66206-25
102095-962EA 20.07 CAD
102095-962 102095-964
Glass Rods, Electron Microscopy Sciences
Stirrers Stirring Rods
Useful tool for all laboratory stirring and mixing
  • 200 mm and 250 mm lengths available
  • Policeman is rubber cover helps help stir at the bottom of container
Useful tool for all laboratories needs to stir/mix substances
Policeman is rubber cover to fit 5 mm rods above. The sealed end is cut at an angle to help stir at the bottom of container.
Glass rods are 5 mm diameter, and come in three lengths.