Swiss Tweezer; Precision, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78170-1G 78170-2AG 78170-2G 78170-3CG 78170-3G 78170-4G 78170-5AG 78170-5ARG 78170-5G 78170-7G
102098-940EA 69.66 CAD
102098-940 102098-942 102098-944 102098-946 102098-948 102098-950 102098-952 102098-954 102098-956 102098-958
Swiss Tweezer; Precision, Electron Microscopy Sciences
Forceps
Swiss precision tweezer made from austenitic, acid-proof stainless steel, 100% anti-magnetic (SA).
  • Tweezers engineered for a NON-SLIP GRIP
  • Light weight –prevents hand fatigue
  • Sensuous touch and functional design
  • Ultra-fine precision tips
  • Melting point 1400°C
Resistant to hydrogen fluoride (HF) and nitric acid.