Micro Scissors, Rubis, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78180-03 78180-1C2 78180-1C3
102098-974EA 109.19 CAD
102098-974 102098-976 102098-978
Micro Scissors, Rubis, Electron Microscopy Sciences
Scissors
Micro scissors manufactured from hardened, anti-acid, stainless, surgical steel (CA)
Rubis Micro Scissors Set includes one light duty Rubis Colibri fine scissors, one medium duty Rubis Classic medium fine scissors, and one heavy duty Rubis Sauro medium strong scissors. Strong, slightly curved easily cut through small bones and cartilages.
Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist® grip scissors offer ergonomically, comfort and control.