Accu-Punch, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69038-02-50 69038-03 69038-03-50 69038-05 69038-05-50 69038-06-50 69038-07 69038-07-50 69038-08 69038-08-50 69038-10 69038-10-50 69038-12 69038-12-50 69038-15 69038-25 69038-35 69038-45 69038-45-50 69038-AS
102096-438EA 573.9 CAD
102096-438 102096-440 102096-442 102096-448 102096-450 102096-454 102096-456 102096-458 102096-460 102096-462 102096-464 102096-466 102096-468 102096-470 102096-472 102096-476 102096-480 102096-484 102096-486 102096-488
Accu-Punch, Electron Microscopy Sciences
Biopsy Punches
The Accu-Punch is a sterile, disposable skin biopsy punch.
  • Always sharp and sterile
  • Ergonomic design
  • Ribbed for maximum control
  • Available in 14 different sizes in a dispenser box
It is the only disposable punch made in the USA.