Single and Multi Wafer Containers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64917-1 64917-2 64917-3 64917-4 64917-46 64917-6 64918-2 64918-3 64918-4
102095-786EA 83.85 CAD
102095-786 102095-788 102095-792 102095-794 102095-796 102095-798 102095-800 102095-802 102095-804
Single and Multi Wafer Containers, Electron Microscopy Sciences
Wafer Handling Supplies Wafer Carriers
These containers are for storage and shipping of a variety of sizes of wafers (Any material) from 1 to 6" in diameter. They prevent damage and contamination to the wafer. Features a conical base to support the wafers on their edges. Wafers are held in place by a spring.
  • Conical base to support the wafers on their edges
  • Wafers are held in place by the spring
The multi-wafer containers are made from polypropylene and have a storage capacity of 25 wafers maximum. They come complete with the base and cover. The single containers will accommodate a variety of sizes from 4 to 6" by simply adjusting the silicone supports in the box. These containers are made from anti-electrostatic polycarbonate and are available in transparent or black.
Delivery information: The set includes the container, the cover and the spring.