Disposable Scalpel Sets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72052-10 72052-11 72052-15 72052-20 72052-22
102097-852EA 26.5 CAD
102097-852 102097-854 102097-856 102097-858 102097-860
Disposable Scalpel Sets, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpels
High quality carbon steel is mounted on plastic handle, individually foil wrapped and sterilized by gamma irradiation.
They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Packaging: Packed 10 each /box.