Cryostat Frozen Sectioning Aid System
Supplier: Electron Microscopy Sciences
62800-X 62800-44
100490-262EA 884.01 CAD
100490-262 100498-218 100498-216 100490-258 100490-260 100490-254 100490-252 100490-250 77548-844
Cryostat Frozen Sectioning Aid System
Microtomes and Microtome Assemblies Microtome Blades and Handles
Frozen sections created as thin as two microns with the same quality as sections cut from paraffin blocks.
  • Preserves sample morphology
  • Increases quality and reduces repeats
  • Fast and effortless serial sectioning
  • Sections can be fixed anhydrously
The CryoJane® tape-transfer system creates frozen sections as thin as 2 µm with the same quality as sections cut from paraffin blocks. Sections are wrinkle-free, uncompressed, and fully intact when bonded to the microscope slide. The Cryogen uses adhesive coated slides and adhesive tapes to capture sections instead of using an anti-roll plate or brush.
With this system, thin tissue sections remain unflawed even after mounting. This makes Cryogen ideal for applications that require perfectly thin sections to clarify structural relationships and reveal morphological detail.
The CryoJane tape-transfer system is adaptable to most cryostats on the market. Sections can be fixed 'anhydrously' to preserve virtually all fine structures present in the snap-frozen block of tissue. Water-soluble enzymes, antigens and nucleo-proteins are preserved in situ. With appropriate fixation and staining, true localization of enzyme and antigen activity can be visualized. The bond between the section and the slide is solvent resistant and permanent, preventing section loss during staining.
CryoJane is suitable for routine cryosections and is indispensable for sectioning difficult tissues including fatty breast and undecalcified bone. Preserves sample morphology. Increases quality, reduces repeats and streamlines workflows. Allows fast and virtually effortless serial sectioning.
Certifications: CE and UL Listed.
Ordering information: The Leica CryoJane tape-transfer system includes the control unit, (ECU) with a power cord, UV flash unit with temperature controlled pad, hand roller and mounting, bracket, door bar, five cable clamps, one slide storage shelf, 125 1X adhesive slides, 10¹/₂X adhesive slides, 10 4X adhesive slides, one roll of tape windows (400 tapes per roll), a manual, one set of replacement fuses, knife facet wipers and a one year parts warranty from date of install.