Willco-dish Kit-Dishes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70655-22 70665-01 70665-02 70665-03 70665-04 70665-05 70665-06 70665-07 70665-08 70665-09 70665-10 70665-11 70665-12 70665-13 70665-15 70665-16 70665-20 70665-21
102097-090EA 644.07 CAD
102097-090 102097-092 102097-094 102097-096 102097-098 102097-100 102097-102 102097-104 102097-106 102097-108 102097-110 102097-112 102097-114 102097-116 102097-120 102097-122 102097-124 102097-126 102097-128
Willco-dish Kit-Dishes, Electron Microscopy Sciences
Culture Dishes Petri Dishes
The WillCo-dish® KIT dishes, of which there are four types in polystyrene (PS) and all four in both transparent as well as in black walled dishes, are designed to offer Scientists the opportunity, to use any chosen material as well as different thicknesses, for the bottom of a dish.
  • Dish (Polystyrene or polycarbonate surrounds and -lids, clean room molded, 500 units each)
  • Pressure sensitive Double Sided [Medical] Adhesive rings, FDA approved (500 units)
  • Glass coverslips: German glass, 0.17 mm. - # 1.5 coverslips (500 units), # 1.5 on request
  • One pair of
More Product Information