Square Petri Dish, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70691 70690-CS 70691-CS
102091-156EA 182.46 CAD
102091-156 102097-176 102097-178
Square Petri Dish, Electron Microscopy Sciences
Culture Dishes Petri Dishes
The square Petri dish with grid is consists of 36 of 13 x 13 mm squares, each 13 mm grid is marked numerically in one direction and alphabetically in the other
Made of polystyrene. Radiation sterile. This dish commonly referred to as a 100 x 15 mm dish. Free from optical distortion and sterile, these dishes are ideal for phase typing, susceptibility testing, plate counts, and probe assays.
The square Petri dish with grid is consists of 36 of 13 x 13 mm squares, each 13 mm grid is marked numerically in one direction and alphabetically in the other. This dish has four venting ribs into the underside of the lid to prevent condensation build-up.
This dish has four venting ribs into the underside of the lid to prevent condensation build-up.