SecurTainer™ I Tamper Evident Specimen Containers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64236-02 64236-04 64236-06 64236-09
102095-006EA 303.93 CAD
102095-006 102095-008 102095-010 102095-012
SecurTainer™ I Tamper Evident Specimen Containers, Electron Microscopy Sciences
Sample Containers General Purpose Containers
Container made of polypropylene and closure made of polyethylene.
  • Unique integrated leak resistant seal
  • High clarity to allow specimen to be viewed without opening
  • The magenta lids are ribbed for easy opening
  • Container are stackable with straight side format
  • SecurTainer™ can be used without using the tamper evident locking mechanism
  • For color coding purpose, use CapInsert
Shatter resistant, especially designed for collection, transport and specimen storage. Both containers and caps are manufactured without the use of plasticizers or mold release agents. All material used are free from latex. All containers are 95 kpa compliant. Non sterile, container and caps are packaged separately in bag of 100.