Petri Dishes — Sterile Petri Dishes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70678-10 70678-20 70678-30 70678-40 70678-50 70678-60 70678-70
102097-162EA 623.8 CAD
102097-162 102097-164 102097-166 102097-168 102097-170 102097-172 102097-174
Petri Dishes — Sterile Petri Dishes, Electron Microscopy Sciences
Culture Dishes Petri Dishes
Made of polystyrene. Our Petri Dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment. Economical, optical clarity, dishes are precision-molded from biomedical grade polystyrene so cultures are clearly visible without distortion. Dishes are packaged 20 each in sturdy polyethylene sleeves. These dishes are not recommended for tissue culture application, due to non treated by “corona effect”.