EMS Grid Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71146-01 71146-02
102097-418EA 28.85 CAD
102097-418 102097-420
EMS Grid Box, Electron Microscopy Sciences
Boxes Storage Boxes
The EMS50 and EMS100 TEM grid storage boxes are used for the storage of TEM grids for routine grid handling, transport and long term TEM grid storage for standard grids that are 3.05 mm in diameter.
  • Dimensions for both boxes are: 75×55×6.5 mm
  • Anti static treated
The EMS 50 and EMS100 capacity inexpensive grid storage boxes. The box has a simple number/letter combination printed on the side of the body.