PetriOrienter and Stand, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70498-09 70498-10 70498-08 70498-07
102097-020EA 40.87 CAD
102097-020 102097-022 102097-024 102097-018
PetriOrienter and Stand, Electron Microscopy Sciences
Well Plate Stands and Well Orienters
Used for working comfortably with solid state media.
  • The Petri Dish Stand: Prevent Fatigues with the 30 angle creates a more comfortable work surface for streaking, transferring, and counting
  • Has unique design, rear slot accepts all PetriOrienters to create a custom layout
The Petri Orienter: Quickly creates a consistent & area of reference on your petri dish. Sharp characters and lines allow for fast and easy position recognition. PetriOrienters are available in several layouts of grids or pie sectors to accommodate colonies of any size.