Sterile Biodegradable Reagent Resevoirs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70497-45 70497-46 70497-47 70497-48
102096-992EA 321.74 CAD
102096-992 102096-994 102096-996 102096-998
Sterile Biodegradable Reagent Resevoirs, Electron Microscopy Sciences
Reagent Reservoirs
Disposable reagent reservoirs.
  • Biodegradable - Using a special additive to PET, these reservoirs will safely degrade over time
  • 100% Eco-Friendly - Each case of Bio-Pure Reservoirs are packed in bio-degradable bags and recycled cardboard cases
A special additive helps the plastic bio-degrade by allowing bacteria and other organisms to breakdown the complex molecular chain. This speeds up the process to reduce the resin to its natural components within five years as opposed to 1,000 years or more.