SecureSeal™ Imaging Spacers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70327-9S 70327-20S 70327-13S 70327-8S
100489-040EA 673.08 CAD
100489-040 100489-038 100501-874 102096-614
SecureSeal™ Imaging Spacers, Electron Microscopy Sciences
Specimen Mounting Supplies
Confine specimens without compression with SecureSeal™ imaging spacers. They are ultra thin adhesive spacers which peel and stick to coverglass or microscope slides to confine specimens without the need for compression.