CultureWell™ Removable Chambered Coverglass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70459-08 70459-08-T
102096-914EA 609.94 CAD
102096-914 103301-916
CultureWell™ Removable Chambered Coverglass, Electron Microscopy Sciences
Cover Glasses
CultureWell™ with removable chambered coverglass is ideal for cell culture and fluorescence imaging applications.
  • Coverslip gasket thickness is 0.5 mm
  • Interior diameter of each well is approx. 6 mm
  • Volume per gasket well without upper structure is approx. 9 µL
  • Volume per well with the upper structure is 250 µL
  • Quality test for non-leakage, cell growth and cover glass removal
The 2 x 8, 16 chambered coverglass is like a chambered slide with the imaging advantages of a removable coverglass substructure
The growth surface is No 1.5 German borosilicate cover glass. Well spacing allows use of multi-channel pipettes for ease and speed of handling.
Black well gasket reduces light scatter, enhancing fluorescence applications. The gasket can easily be removed from the cover glass for mounting. Gasket well design is ideally suited for small volume incubations, in-situ hybridization and immunostaining.
Silicone well gasket remains attached to cover glass after separation, allowing wells to be used as reagent reservoirs.
Ordering information: Frosted microscope slides are provided for mounting.