ProPlate™ MicroArray System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63484-81 63484-15 63484-82 63484-58 63484-21 63484-03 63484-24 63484-57 63484-36 63484-23 63484-47 63484-26 63484-05 63484-45 63484-55 63484-46 63484-10 63484-06 63484-27 63484-13 63484-04 63484-37 63484-31 63484-35 63484-32 63484-54 63484-83 63484-22 63484-51 63484-01 63484-38 63484-42 63484-28 63484-84 63484-33 63484-11 63484-43 63484-48 63484-14 63484-52 63484-53 63484-25 63484-56 63484-44 63484-41 63484-07 63484-09 63484-12 63484-02 63484-08 63484-16 63484-34 63484-80
102094-630EA 560.02 CAD
102094-630 102094-674 102094-672 102094-670 102094-708 102094-706 102094-628 102094-704 102094-626 102094-702 102094-668 102094-624 102094-700 102094-666 102094-622 102094-664 102094-662 102094-660 102094-658 102094-656 102094-654 102094-698 102094-696 102094-652 102094-650 102094-694 102094-692 102094-690 102094-726 102094-648 102094-724 102094-646 102094-722 102094-688 102094-644 102094-720 102094-642 102094-686 102094-640 102094-684 102094-682 102094-680 102094-718 102094-716 102094-638 102094-714 102094-636 102094-712 102094-678 102094-634 102094-710 102094-632 102094-676
ProPlate™ MicroArray System, Electron Microscopy Sciences
Microarray Instruments
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.