Labeling Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77627-B 77627-G 77627-R 77627-W 77627-Y 77628-B 77628-G 77628-R 77628-W 77628-Y
102098-756EA 34.03 CAD
102098-756 102098-758 102098-760 102098-762 102098-764 102098-766 102098-768 102098-770 102098-772 102098-774
Labeling Tape, Electron Microscopy Sciences
Tapes
Use pen, pencil or marker to mark. Leaves no adhesive residue when peeled off.
Water, oil, and acid resistant. Withstands cold (−23 °C) and hot (121 °C) temperatures.