Write Protect Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77626-10 77626-20
102098-752EA 77.53 CAD
102098-752 102098-754
Write Protect Tape, Electron Microscopy Sciences
Tapes
Write Protect is a clear adhesive tape designed to cover and protect writing or printing on labels, on the surface of microtubes and on other laboratory containers.
  • Will not dry out, peel or fall off
  • Withstand temperatures ranging from –196 to 150 °C
The clear tape comes in a handy dispenser and will adhere to all plastics, glass, metals and label materials. The tape is provided in perforated, pull-off segments for 0.5 and 1.5 mL tubes.