Glow Writer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68551-01 68551-RT
102096-328EA 25.53 CAD
102096-328 102096-326
Glow Writer, Electron Microscopy Sciences
Pens Autoradiography Pens
Fine-point phosphorescent marking pen for laboratory autography.
Glow Writer is a phosphorescent pen for marking chemiluminescent and isotope-labeled substrates for X-ray film autography. The unique marker delivers a phosphor-containing ink through a fine point porous nib. Before exposure to X-ray film, the phosphorescent writing is excited by room light. The pen utilizes a phosphor ink having an optimum concentration of phosphor and a relatively short afterglow half-life (approximately 2-3 minutes) has been selected so that upon exposure to X-ray film, autographic exposures of almost any duration will be easily readable on the film. The fine point nib is easily replaceable if it dries out.