Rota-Rack Rotating Tube Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61604-10 61604-20
102093-674EA 170.07 CAD
102093-674 102093-676
Rota-Rack Rotating Tube Rack, Electron Microscopy Sciences
Racks Tube Racks
Each module of the Rota-Rack for large tubes holds two 50 mL tubes, eight 15 mL tubes, six 20 mm tubes, or ten 10-13 mm tubes
Connect up to 4 interlocking racks. Twist and turn to suit your application. Rotate each of the four modules independently. Use several sizes of test tubes, culture tubes, centrifuge tubes, or microcentrifuge tubes at the same time. Click each module into place, load with tubes, and carry. Rack maintains its orientation until you twist it again. Comes fully assembled. Autoclavable.
Each module of the Rota-Rack for large tubes holds two 50 mL tubes, eight 15 mL tubes, six 20 mm tubes, or ten 10-13 mm tubes. Each module of the Rota Rack for microtubes holds six 15 mL tubes, nine 1.5/2.0 mL tubes, twelve 0.5/0.6 mL tubes, or thirty-two 0.2 mL PCR* tubes or four 8-tube strips.
Each module of the Rota Rack for microtubes holds six 15 mL tubes, nine 1.5/2.0 mL tubes, twelve 0.5/0.6 mL tubes, or thirty-two 0.2 mL PCR* tubes or four 8-tube strips.