81-Well Storage Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61600-A 61600-B 61600-G 61600-N 61600-O 61600-P 61600-Y
102093-618EA 141.3 CAD
102093-618 102093-620 102093-622 102093-624 102093-626 102093-628 102093-630
81-Well Storage Rack, Electron Microscopy Sciences
Boxes Boxes for Tubes
Durable polypropylene rack fits in standard 5.25” square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials.
  • Molded grid lines on the lid
  • Imprinted coordinates on the bottom of base
  • Molded reference point on the lid
  • Rack measures 5.125×5.125×1.75 cm