Media Disruptor Genie, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72408-01 72407-20 72408-05
101454-154EA 107.12 CAD
101454-154 101454-144 101454-156
Media Disruptor Genie, Electron Microscopy Sciences
Homogenizers Ultrasonic Cell Disruptors
Rapid hands-free disruption of up to 12 microtubes (1.5 ml or 2.0 ml) at one time.
  • Unique agitation, vortexing, and collision actions for cell disruption and sample resuspension
  • Maintain operating speed for up to 15 minutes or continuous operation, even in cold rooms
  • Supplied pop-up cup easily attaches for use as a standard single-tube vortex mixer
  • Ideal for glass bead procedures using Disruptor Beads
  • Less costly than ultrasonic units or homogenizes
  • Spherecal lead-free soda lime glass disruptor beads are available in two sizes
  • Beads can be used for disruption of yeast, bacteria, and other samples
  • Beads can be used in combination with Disruptor Genie or TurboMix™ attachments for Vortex-Genie™ 2, Vortec-Genie™ 2T or Digital Vortex-Genie™ 2