Glass Bead Sterilizers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66117-10 66117-20 66116-5 66117-3 66117-5 66116-20 66116-10
102095-938EA 309.59 CAD
102095-936 102095-928 102095-938 102095-934 102095-940 102095-942 102095-930
Glass Bead Sterilizers, Electron Microscopy Sciences
Sterilizers Glass Bead Sterilizers
Glass beads are suggested to be replaced at least once in three months
These glass bead sterilizers are quick and easy to use and they are compact and light weight which makes them suitable for table tops or workbenches. They are used to sterilize instruments used in most laboratories and doctors' offices, dentists' offices, clinics, hospitals etc, killing  spores and bacteria within 10 seconds. All units are supplied with one filling of glass beads. Glass beads size ranging 1.5 – 2.0 mm diameter.
Glass beads are suggested to be replaced at least once in three months. Clean the glass beads at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
*Model BS300 is available in colors: Black, Red, Green, Brown, Blue and Gray. Please indicate your color when ordering. If you’re not mentioned your color, we automatically ship color Blue.
*Model BS300 is available in colors: Black, Red, Green, Brown, Blue and Gray. Please indicate your color when ordering. If you’re not mentioned your color, we automatically ship color Blue.
Clean the glass beads at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.