Cryogenic Grid Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71166-10 71166-20 71166-30
102097-442EA 29.83 CAD
102097-442 102097-444 102097-446
Cryogenic Grid Box, Electron Microscopy Sciences
Boxes Cryoboxes
These specimen grid boxes are for storing or transferring cryogenic TEM specimen grids.
  • Four diamond shaped slots
  • Non-static clear cover shield held in place with stainless steel screw, which is tapped in the center of the box
  • Box only fits 71166-20 (Grid box square with lid)
  • Available with lid, pin-type lid, or without lid
Order Now
SPECIFICATIONS
Environmentally Preferable