Stainless Steel Brain and Heart Matrices, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69090-C 69090-S 69091-C 69091-S 69092-C 69092-S 69095-H 69096-H
102096-502EA 1175.19 CAD
102096-502 102096-504 102096-506 102096-508 102096-510 102096-512 102096-514 102096-516
Stainless Steel Brain and Heart Matrices, Electron Microscopy Sciences
Specimen Mounting Supplies
When your rodent brain, heart, or other similar organ needs to be sliced as thinly as possible, these new stainless steel matrices with a 0.5mm channel will achieve these slices – coronal or Sagittal – with ease.
  • Made of high-quality stainless steel
  • Can be repeatedly used, frozen, autoclaved, and cleaned
  • Produces fine slices
  • Accurate sectioning
  • Easy cleaning and easy handling